1,6-Diaminohexan ≥70%, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,6-Hexandiamin, Hexamethylendiamin

M-Clarity™ Qualitätsstufe = MQ200

422002-2.5L
SIAL422002-2.5LEA 127 EUR
SIAL422002-2.5L
1,6-Diaminohexan ≥70%, Sigma-Aldrich®
1,6-Diaminohexan
Formel: C₆H₁₆N₂
Molecular Weight: 116,21 g/mol
MDL: MFCD00008243
CAS-Nummer: 124-09-4
EINECS: 204-679-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR