2,6-Dichlor-5-fluornicotinonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

422169-25G
SIAL422169-25GEA 121 EUR
SIAL422169-25G
2,6-Dichlor-5-fluornicotinonitril, Sigma-Aldrich®
Formel: C₆HCl₂FN₂
MDL: MFCD01863289
CAS-Nummer: 82671-02-1
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR