4-(Trifluormethoxy)benzamidoxim, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Hydroxy-4-(trifluoromethoxy)benzimidamide

M-Clarity™ Qualitätsstufe = MQ100

422231-1G
SIAL422231-1GEA 38.7 EUR
SIAL422231-1G
4-(Trifluormethoxy)benzamidoxim, Sigma-Aldrich®
4-(Trifluormethoxy)benzamidoxim
Formel: C₈H₇F₃N₂O₂
Molecular Weight: 220,15 g/mol
MDL: MFCD00085175
CAS-Nummer: 56935-71-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR