Thiazol-2-carbaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Thiazolecarboxaldehyde, Thiazole-2-carboxaldehyde

M-Clarity™ Qualitätsstufe = MQ100

422460-1G
SIAL422460-1GEA 78.8 EUR
SIAL422460-1G
Thiazol-2-carbaldehyd, Sigma-Aldrich®
Thiazol-2-carbaldehyd
Formel: C₄H₃NOS
Molecular Weight: 113.13 g/mol
MDL: MFCD00142924
CAS-Nummer: 10200-59-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR