1-Butyl-3-methylimidazoliumthiocyanat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-butyl-3-methylimidazolium thiocyanate

42254-100G-F
SIAL42254-100G-FEA 743 EUR
SIAL42254-100G-F
1-Butyl-3-methylimidazoliumthiocyanat, Sigma-Aldrich®
1-Butyl-3-methylimidazolium thiocyanate
Formel: C₉H₁₅N₃S
Molecular Weight: 197.3 g/mol
MDL: MFCD06798181
CAS-Nummer: 344790-87-0
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR