(R)-(+)-2-Pyrrolidinon-5-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: D-Pyr-OH, (R)-(+)-2-Pyrrolidone-5-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

422614-10G
SIAL422614-10GEA 102 EUR
SIAL422614-10G
(R)-(+)-2-Pyrrolidinon-5-carbonsäure, Sigma-Aldrich®
(R)-(+)-2-Pyrrolidinon-5-carbonsäure
Formel: C₅H₇NO₃
Molecular Weight: 129,12 g/mol
MDL: MFCD00066212
CAS-Nummer: 4042-36-8
EINECS: 223-735-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR