4-Nitro-o-anissäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ200

422916-5G
SIAL422916-5GEA 53.2 EUR
SIAL422916-5G
4-Nitro-o-anissäure, Sigma-Aldrich®
4-Nitro-o-anissäure
Formel: C₈H₇NO₅
Molecular Weight: 197,15 g/mol
MDL: MFCD00216573
CAS-Nummer: 2597-56-0
EINECS: 219-998-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR