(Hydroxypropyl)methylcellulose (Hypromellose), M.W. ~120,000, 100,000 mPa.s (2% in H2O), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

423173-100G
SIAL423173-100GEA 141 EUR
SIAL423173-100G
(Hydroxypropyl)methylcellulose (Hypromellose), M.W. ~120,000, 100,000 mPa.s (2% in H2O), Sigma-Aldrich®
(Hydroxypropyl)methylcellulose (Hypromellose)
Formel: C₃H₈O₂·xCH₄O
MDL: MFCD00131360
CAS-Nummer: 9004-65-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR