3,5-Dimethoxytoluol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 1,3-Dimethoxy-5-methylbenzol

M-Clarity™ Qualitätsstufe = MQ100

423742-1G
SIAL423742-1GEA 44.4 EUR
SIAL423742-1G
3,5-Dimethoxytoluol, Sigma-Aldrich®
3,5-Dimethoxytoluol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
MDL: MFCD00015435
CAS-Nummer: 4179-19-5
EINECS: 224-048-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR