3-Aminoisoxazol, Sigma-Aldrich®

Lieferant: Merck


MQ100

424218-25ML 424218-5ML
SIAL424218-25MLEA 444 EUR
SIAL424218-25ML SIAL424218-5ML
3-Aminoisoxazol, Sigma-Aldrich®
3-Aminoisoxazole
Formel: C₃H₄N₂O
Molecular Weight: 84.08 g/mol
MDL: MFCD00038814
CAS-Nummer: 1750-42-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR