1,2,3,4-Tetramethylcyclopentadien, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

424471-5G
SIAL424471-5GEA 271 EUR
SIAL424471-5G
1,2,3,4-Tetramethylcyclopentadien, Sigma-Aldrich®
1,2,3,4-Tetramethylcyclopentadien
Formel: C₉H₁₄
MDL: MFCD00145385
CAS-Nummer: 4249-10-9
UN: 3295
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR