(4-Chlorbenzyl)triphenylphosphoniumchlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

424587-5G
SIAL424587-5GEA 49.1 EUR
SIAL424587-5G
(4-Chlorbenzyl)triphenylphosphoniumchlorid, Sigma-Aldrich®
(4-Chlorbenzyl)triphenylphosphoniumchlorid
Formel: C₂₅H₂₁Cl₂P
Molecular Weight: 423,32 g/mol
MDL: MFCD00041533
CAS-Nummer: 1530-39-8
EINECS: 216-228-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR