trans-2-Nitro-1-phenylpropen, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

424749-5G
SIAL424749-5GEA 83.5 EUR
SIAL424749-5G
trans-2-Nitro-1-phenylpropen, Sigma-Aldrich®
trans-2-Nitro-1-phenylpropen
Formel: C₉H₉NO₂
CAS-Nummer: 18315-84-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR