trans-4-Methyl-β-nitrostyrol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (E)-1-Methyl-4-(2-nitrovinyl)benzene

M-Clarity™ Qualitätsstufe = MQ100

424757-5G
SIAL424757-5GEA 93.3 EUR
SIAL424757-5G
trans-4-Methyl-β-nitrostyrol, Sigma-Aldrich®
trans-4-Methyl-β-nitrostyrol
Formel: C₉H₉NO₂
MDL: MFCD00024828
CAS-Nummer: 7559-36-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR