1,3,5-Tris(trifluormethyl)benzol, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
424765-1G
424765-5G
SIAL424765-1GEA
88.2
EUR
SIAL424765-1G
SIAL424765-5G
1,3,5-Tris(trifluormethyl)benzol, Sigma-Aldrich®
1,3,5-Tris(trifluormethyl)benzol
Formel:
C₉H₃F₉ Molecular Weight: 282.105 g/mol |
MDL:
MFCD00054722 CAS-Nummer: 729-81-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...