Dysprosium(III)trifluormethansulfonat, Sigma-Aldrich®

Lieferant: Merck

425664-5G
SIAL425664-5GEA 167 EUR
SIAL425664-5G
Dysprosium(III)trifluormethansulfonat, Sigma-Aldrich®
Dysprosium(III)trifluormethansulfonat
Formel: C₃DyF₉O₉S₃
MDL: MFCD00209583
CAS-Nummer: 139177-62-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR