Holmium(III)trifluormethansulfonat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

425699-1G
SIAL425699-1GEA 48.6 EUR
SIAL425699-1G
Holmium(III)trifluormethansulfonat, Sigma-Aldrich®
Holmium(III)trifluormethansulfonat
Formel: C₃F₉HoO₉S₃
Molecular Weight: 612,14 g/mol
CAS-Nummer: 139177-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR