Samarium(III)trifluormethansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Samarium(III) trifluoromethanesulphonate

425710-10G 425710-1G
SIAL425710-10GEA 99.6 EUR
SIAL425710-10G SIAL425710-1G
Samarium(III)trifluormethansulfonat, Sigma-Aldrich®
Samarium(III)trifluormethansulfonat
Formel: C₃F₉O₉S₃Sm
MDL: MFCD00192431
CAS-Nummer: 52093-28-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR