Thulium(III)trifluormethansulfonat ≥98%, Sigma-Aldrich®

Lieferant: Merck

425737-1G
SIAL425737-1GEA 125 EUR
SIAL425737-1G
Thulium(III)trifluormethansulfonat ≥98%, Sigma-Aldrich®
Thulium(III)trifluormethansulfonat
Formel: C₃F₉O₉S₃Tm
Molecular Weight: 616,14 g/mol
MDL: MFCD00209617
CAS-Nummer: 141478-68-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR