Methyl-2,5-dihydroxybenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,5-Dihydroxybenzoesäuremethylester, Gentisinsäuremethylester

426091-25G
SIAL426091-25GEA 429 EUR
SIAL426091-25G
Methyl-2,5-dihydroxybenzoat, Sigma-Aldrich®
Methyl-2,5-dihydroxybenzoat
Formel: C₈H₈O₄
Molecular Weight: 168,15 g/mol
MDL: MFCD00016464
CAS-Nummer: 2150-46-1
EINECS: 218-427-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR