Tetraisopropylmethylenbisphosphonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetraisopropyl methylenebis(phosphonate), Tetraisopropyl methylenediphosphonate

M-Clarity™ Qualitätsstufe = MQ100

426431-25ML
SIAL426431-25MLEA 148 EUR
SIAL426431-25ML
Tetraisopropylmethylenbisphosphonat, Sigma-Aldrich®
Tetraisopropylmethylenbisphosphonat
Formel: C₁₃H₃₀O₆P₂
Molecular Weight: 344.33 g/mol
MDL: MFCD00015021
CAS-Nummer: 1660-95-3
EINECS: 216-765-0
UN: 3278
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR