Poly(ethylene-co-acrylic acid), Perlen, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

426717-250G
SIAL426717-250GEA 65.6 EUR
SIAL426717-250G
Poly(ethylene-co-acrylic acid), Perlen, Sigma-Aldrich®
Poly(ethylene-co-acrylic acid)

Used as processing and performance additive. Promotes crystallisation of PET. Assists dispersion of additives in plastics.


Forms reversible ionic clusters (crosslinks). Promotes adhesion to various substrates, tougher, more chemically resistant and more transparent than parent acid copolymer.

Formel: C₁₉H₁₉N₃O₆
Dichte: 0,93 g/ml (25 °C)
MDL: MFCD00133995
CAS-Nummer: 9010-77-9
UN: 3261
ADR: 8,I

Bestellen

Spezifikation Testergebnisse

Infrared Spectrum Conforms to Structure
Acid Value 37 - 44 mg/g
Transition Temperature 99 - 101 °C
Viscosity 575 cP (145 °C, Brookfield)(lit.)
Drop Point 105 °C, Mettler
Hardness ≤2.5 dmm (ASTM D 5, 25 °C)
Composition Acrylic Acid, 5 wt.%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR