(R)-3-(Boc-amino)-3-(3-methylphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-3-((tert-Butoxycarbonyl)amino)-3-(m-tolyl)propanoic acid

42722-500MG-F
SIAL42722-500MG-FEA 354 EUR
SIAL42722-500MG-F
(R)-3-(Boc-amino)-3-(3-methylphenyl)propionsäure, Sigma-Aldrich®
(R)-3-(Boc-amino)-3-(3-methylphenyl)propionsäure
Formel: C₁₅H₂₁NO₄
Molecular Weight: 279,3315 g/mol
MDL: MFCD03427934
CAS-Nummer: 464930-76-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR