Dimethoxy(diphenyl)silan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diphenyldimethoxysilan

42940-100ML
SIAL42940-100MLEA 115 EUR
SIAL42940-100ML
Dimethoxy(diphenyl)silan, Sigma-Aldrich®
Dimethoxy(diphenyl)silan
Formel: C₁₄H₁₆O₂Si
Molecular Weight: 244,37 g/mol
MDL: MFCD00025690
CAS-Nummer: 6843-66-9
EINECS: 229-929-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR