(R)-(+)-2-Brom-α-methylbenzylalkohol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-(+)-1-(2-Bromophenyl)ethanol, (R)-2-Bromo-alpha-methylbenzyl Alcohol

M-Clarity™ Qualitätsstufe = MQ100

429791-5G
SIAL429791-5GEA 612 EUR
SIAL429791-5G
(R)-(+)-2-Brom-α-methylbenzylalkohol, Sigma-Aldrich®
(R)-(+)-2-Brom-α-methylbenzylalkohol
Formel: C₈H₉BrO
Molecular Weight: 201,06 g/mol
MDL: MFCD00216591
CAS-Nummer: 76116-20-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR