2,5-Dichlor-3-nitrobenzoesäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

429848-5G
SIAL429848-5GEA 508 EUR
SIAL429848-5G
2,5-Dichlor-3-nitrobenzoesäure, Sigma-Aldrich®
2,5-Dichloro-3-nitrobenzoic acid
Formel: C₇H₃Cl₂NO₄
Molecular Weight: 236,01 g/mol
MDL: MFCD00014691
CAS-Nummer: 88-86-8
EINECS: 201-862-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR