(Heptafluorpropyl)trimethylsilan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Trimethyl(heptafluoropropyl)silane, Trimethyl(perfluoropropyl)silane
M-Clarity™ Qualitätsstufe = MQ100
431044-1G
SIAL431044-1GEA
116
EUR
SIAL431044-1G
(Heptafluorpropyl)trimethylsilan, Sigma-Aldrich®
(Heptafluoropropyl)trimethylsilane
Formel:
C₆H₉F₇Si |
MDL:
MFCD00216712 CAS-Nummer: 3834-42-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...