Vitamin B12 (Cyanocobalamin), Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
43107-10G-F
SIAL43107-10G-FEA
1160
EUR
SIAL43107-10G-F
Vitamin B12 (Cyanocobalamin), Sigma-Aldrich®
Vitamin B12 (Cyanocobalamin)
Formel:
C₆₃H₈₈CoN₁₄O₁₄P Molecular Weight: 1355,38 g/mol Schmelzpunkt: 320 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00151092 CAS-Nummer: 68-19-9 EINECS: 200-680-0 Merck Index: 12,10152 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...