α,α-Diphenyl-N-methyl-D-prolinol, Sigma-Aldrich®

Lieferant: Merck

43118-250MG
SIAL43118-250MGEA 71.4 EUR
SIAL43118-250MG
α,α-Diphenyl-N-methyl-D-prolinol, Sigma-Aldrich®
α,α-Diphenyl-N-methyl-D-prolinol
Formel: C₁₈H₂₁NO
MDL: MFCD00145217
CAS-Nummer: 144119-12-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR