2,5-Dichlor-p-benzochinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,5-Dichloro-1,4-benzoquinone, 2,5-Dichlorobenzo-1,4-quinone

M-Clarity™ Qualitätsstufe = MQ200

431974-10G
SIAL431974-10GEA 208 EUR
SIAL431974-10G
2,5-Dichlor-p-benzochinon, Sigma-Aldrich®
2,5-Dichlor-p-benzochinon
Formel: C₆H₂Cl₂O₂
Molecular Weight: 176.99 g/mol
MDL: MFCD00041736
CAS-Nummer: 615-93-0
EINECS: 210-453-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR