2,6-Dichlor-p-benzochinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,6-Dichloro-1,4-benzoquinone

M-Clarity™ Qualitätsstufe = MQ200

431982-5G
SIAL431982-5GEA 92.2 EUR
SIAL431982-5G
2,6-Dichlor-p-benzochinon, Sigma-Aldrich®
2,6-Dichlor-p-benzochinon
Formel: C₆H₂Cl₂O₂
Molecular Weight: 176,99 g/mol
MDL: MFCD00037159
CAS-Nummer: 697-91-6
EINECS: 211-810-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR