Methyl-2,6-dihydroxybenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,6-Dihydroxybenzoesäuremethylester

432792-1G
SIAL432792-1GEA 32.3 EUR
SIAL432792-1G
Methyl-2,6-dihydroxybenzoat, Sigma-Aldrich®
Methyl-2,6-dihydroxybenzoat
Formel: C₈H₈O₄
Molecular Weight: 168,15 g/mol
MDL: MFCD00059619
CAS-Nummer: 2150-45-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR