1,3-Didecyl-2-methylimidazoliumchlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

433780-1G
SIAL433780-1GEA 59.3 EUR
SIAL433780-1G
1,3-Didecyl-2-methylimidazoliumchlorid, Sigma-Aldrich®
1,3-Didecyl-2-methylimidazolium chloride
Formel: C₂₄H₄₇ClN₂
CAS-Nummer: 70862-65-6
EINECS: 274-948-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR