1-Decyl-2-methylimidazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Decyl-2-methyl-1H-imidazol, N-Decyl-2-methylimidazol, N-Decyl-2-methyl-1H-imidazol
MQ100
433799-25ML
SIAL433799-25MLEA
103
EUR
SIAL433799-25ML
1-Decyl-2-methylimidazol, Sigma-Aldrich®
1-Decyl-2-methylimidazole
Formel:
C₁₄H₂₆N₂ Molecular Weight: 222.37 g/mol |
CAS-Nummer:
42032-30-4 EINECS: 255-633-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...