5-Chlor-3-methyl-1-phenyl-1H-pyrazol-4-carbaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carboxaldehyde

M-Clarity™ Qualitätsstufe = MQ100

433853-5G
SIAL433853-5GEA 255 EUR
SIAL433853-5G
5-Chlor-3-methyl-1-phenyl-1H-pyrazol-4-carbaldehyd, Sigma-Aldrich®
5-Chlor-3-methyl-1-phenyl-1H-pyrazol-4-carbaldehyd
Formel: C₉H₁₁ClN₂O
Molecular Weight: 220,65 g/mol
MDL: MFCD00100755
CAS-Nummer: 947-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR