Eisen(II)bromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Eisendibromid

M-Clarity™ Qualitätsstufe = MQ100

434000-1G
SIAL434000-1GEA 66.8 EUR
SIAL434000-1G
Eisen(II)bromid, Sigma-Aldrich®
Eisen(II)bromid
Formel: Br₂Fe
Molecular Weight: 215,66 g/mol
MDL: MFCD00016081
CAS-Nummer: 7789-46-0
EINECS: 232-168-8
UN: 3260
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR