2-Hydroxy-3-methoxy-5-nitrobenzaldehyd, Sigma-Aldrich®

Lieferant: Merck

434086-5G
SIAL434086-5GEA 162 EUR
SIAL434086-5G
2-Hydroxy-3-methoxy-5-nitrobenzaldehyd, Sigma-Aldrich®
2-Hydroxy-3-methoxy-5-nitrobenzaldehyde
Formel: C₈H₇NO₅
Molecular Weight: 197,15 g/mol
MDL: MFCD00017033
CAS-Nummer: 17028-61-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR