Tetraethylammoniumhexafluorophosphat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetraethylammoniumhexafluorophosphate

M-Clarity™ Qualitätsstufe = MQ200

434116-25G
SIAL434116-25GEA 187 EUR
SIAL434116-25G
Tetraethylammoniumhexafluorophosphat, Sigma-Aldrich®
Tetraethylammonium hexafluorophosphate
Formel: C₈H₂₀F₆NP
MDL: MFCD00043179
CAS-Nummer: 429-07-2
EINECS: 207-056-7
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR