Bortrifluorid-Tetrahydrofuran-Komplex, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

434280-100ML 434280-500ML
SIAL434280-100MLEA 43.4 EUR
SIAL434280-100ML SIAL434280-500ML
Bortrifluorid-Tetrahydrofuran-Komplex, Sigma-Aldrich®
Bortrifluorid-Tetrahydrofuran-Komplex
Formel: C₄H₈BF₃O
MDL: MFCD00040372
CAS-Nummer: 462-34-0
EINECS: 207-325-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR