(S)-(+)-2,2-Dimethylcyclopropancarbonsäureamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-2,2-Dimethylcyclopropanecarboxamide

M-Clarity™ Qualitätsstufe = MQ100

434639-10G
SIAL434639-10GEA 89.9 EUR
SIAL434639-10G
(S)-(+)-2,2-Dimethylcyclopropancarbonsäureamid, Sigma-Aldrich®
(S)-(+)-2,2-Dimethylcyclopropancarbonsäureamid
Formel: C₆H₁₁NO
Molecular Weight: 113,1600 g/mol
MDL: MFCD00216614
CAS-Nummer: 75885-58-4
EINECS: 278-334-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR