2-(Benzyloxy)-3-methoxybenzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

434795-1G
SIAL434795-1GEA 92.3 EUR
SIAL434795-1G
2-(Benzyloxy)-3-methoxybenzaldehyd, Sigma-Aldrich®
2-(Benzyloxy)-3-methoxybenzaldehyd
Formel: C₁₅H₁₄O₃
MDL: MFCD00202674
CAS-Nummer: 2011-06-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR