Methyl hydroxyethylcellulose, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

435015-250G
SIAL435015-250GEA 44.1 EUR
SIAL435015-250G
Methyl hydroxyethylcellulose, Sigma-Aldrich®
Methyl hydroxyethylcellulose

Water retention aid, thickening agent, protective colloid, suspending agent, binder and stabiliser.


Extent of labeling varies from 0.06 to 0.50 mol hydroxyethyl per mol cellulose and 1.3 to 2.2 mol methyl per mol cellulose. It has 8% (by weight) of hydroxyethyl and 26% (by weight) of methoxy groups.

MDL: MFCD00147597
CAS-Nummer: 9032-42-2

Bestellen

Spezifikation Testergebnisse

Infrared Spectrum Conforms to Structure
Viscosity (c = 2%, Water, 20 °C) 15000 - 20500 cps
Water (by Karl Fischer) as Packaged ≤8.00%
Particle Size (µm), Laser DV90 150 - 250


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR