Pluronic® L-35 (Synperonic® L 35), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Synperonic® L35, PEG-PPG-PEG, Pluronic® L-35

M-Clarity™ Qualitätsstufe = MQ100

435414-250ML
SIAL435414-250MLEA 92.8 EUR
SIAL435414-250ML
Pluronic® L-35 (Synperonic® L 35), Sigma-Aldrich®
Pluronic® L-35 (Synperonic® L 35)

Hard and soft surface cleaners, defoamers in coatings and water treatment. Lubricant in metal working, anti-foaming aid and extender for linear and cross-linked polyesters and polyurethanes.

Molecular Weight: 1900 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 9003-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR