Pluronic® L-61 Tensid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: PEG-PPG-PEG, Synperonic® L61

M-Clarity™ Qualitätsstufe = MQ200

435422-250ML
SIAL435422-250MLEA 102 EUR
SIAL435422-250ML
Pluronic® L-61 Tensid, Sigma-Aldrich®
Pluronic® L-61 (Synperonic® L 61)

Hard and soft surface cleaners, defoamers in coatings and water treatment. Lubricant in metal working, anti-foaming aid and extender for linear and cross-linked polyesters and polyurethanes.

Formel: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
Molecular Weight: 2000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00082049
CAS-Nummer: 9003-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR