(S)-(+)-Carvon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
D(+)-Carvon, (S)-(+)-2-Methyl-5-(1-methylvinyl)cyclohex-2-en-1-on, D-p-Mentha-1(6),8-dien-2-on
M-Clarity™ Qualitätsstufe = MQ100
435759-25ML
435759-5ML
SIAL435759-25MLEA
121
EUR
SIAL435759-25ML
SIAL435759-5ML
(S)-(+)-Carvon, Sigma-Aldrich®
(S)-(+)-Carvon
Formel:
C₁₀H₁₄O Molecular Weight: 150.22 g/mol Siedepunkt: 230 Dichte: 0.9584 g/cm³ (20 °C) |
MDL:
MFCD00062997 CAS-Nummer: 2244-16-8 EINECS: 218-827-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...