Indium(III)fluorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
435848-1G
SIAL435848-1GEA
57.7
EUR
SIAL435848-1G
Indium(III)fluorid, Sigma-Aldrich®
Indium(III)fluorid
Formel:
F₃In Schmelzpunkt: 1170 °C |
MDL:
MFCD00016149 CAS-Nummer: 7783-52-0 EINECS: 232-005-0 UN: 2923 ADR: 8,II Merck Index: 12,04990 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...