Eisen(III)phosphat Tetrahydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Eisen(III)orthophosphat Tetrahydrat, Eisenphosphat Tetrahydrat

M-Clarity™ Qualitätsstufe = MQ200

436038-100G
SIAL436038-100GEA 36.7 EUR
SIAL436038-100G
Eisen(III)phosphat Tetrahydrat, Sigma-Aldrich®
Eisen(III)phosphat Tetrahydrat
Formel: FeO₄P·4H₂O
Molecular Weight: 222,88 g/mol
CAS-Nummer: 31096-47-6
EINECS: 233-149-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR