2-Fluor-3-(trifluormethyl)phenol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Fluoro-3-hydroxybenzotrifluoride

M-Clarity™ Qualitätsstufe = MQ100

436119-5G
SIAL436119-5GEA 119 EUR
SIAL436119-5G
2-Fluor-3-(trifluormethyl)phenol, Sigma-Aldrich®
2-Fluor-3-(trifluormethyl)phenol
Formel: C₇H₄F₄O
Molecular Weight: 180,1 g/mol
MDL: MFCD00083554
CAS-Nummer: 207291-85-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR