4-Chlor-2-fluorbenzoesäure, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
436763-5G
SIAL436763-5GEA
47.5
EUR
SIAL436763-5G
4-Chlor-2-fluorbenzoesäure, Sigma-Aldrich®
4-Chloro-2-fluorobenzoic acid
Formel:
C₇H₄ClFO₂ Molecular Weight: 174,56 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042468 CAS-Nummer: 446-30-0 EINECS: 207-162-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...