1-(Methoxymethyl)benzotriazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(Methoxymethyl)-1H-benzotriazole

M-Clarity™ Qualitätsstufe = MQ100

438022-1G
SIAL438022-1GEA 106 EUR
SIAL438022-1G
1-(Methoxymethyl)benzotriazol, Sigma-Aldrich®
1-(Methoxymethyl)benzotriazole
Formel: C₈H₉N₃O
MDL: MFCD00216630
CAS-Nummer: 71878-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR